Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD109 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159587.2 | NP_001153059.1 | 25 | 3134 | ||
NM_001159588.2 | NP_001153060.1 | 24 | 2903 | |||
NM_133493.4 | NP_598000.2 | 25 | 3134 | |||
XM_005248659.3 | XP_005248716.1 | 26 | 3281 | |||
XM_011535473.1 | XP_011533775.1 | 25 | 2979 | |||
XM_017010283.1 | XP_016865772.1 | 23 | 2746 | |||
GenBank mRNA | AF410459.1 | - | 25 | 3115 | ||
AK123960.1 | - | 11 | 1212 | |||
AK313636.1 | - | 25 | 3114 | |||
AL834478.1 | - | 11 | 1230 | |||
AY149920.1 | - | 25 | 3003 | |||
AY788891.1 | - | 25 | 3028 | |||
BX641095.1 | - | 24 | 2790 | |||
EF553520.1 | - | 25 | 3033 |