Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAET1E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243325.1 | NP_001230254.1 | 3, 4 | 649 | ||
NM_139165.2 | NP_631904.1 | 3, 4 | 757 | |||
XM_011535475.2 | XP_011533777.1 | 5, 6 | 1277 | |||
XM_011535476.2 | XP_011533778.1 | 5, 6 | 1291 | |||
XM_011535478.2 | XP_011533780.1 | 5, 6 | 1274 | |||
XM_017010285.1 | XP_016865774.1 | 5, 6 | 1276 | |||
XM_017010286.1 | XP_016865775.1 | 5, 6 | 1276 | |||
XM_017010287.1 | XP_016865776.1 | 5, 6 | 1276 | |||
XM_017010288.1 | XP_016865777.1 | 3, 4 | 525 | |||
GenBank mRNA | AF359243.1 | - | 3, 4 | 645 | ||
AY054974.1 | - | 3, 4 | 516 | |||
AY069961.1 | - | 3, 4 | 684 | |||
AY176317.1 | - | 3, 4 | 624 | |||
AY252119.1 | - | 3, 4 | 624 | |||
BC101976.1 | - | 3, 4 | 549 | |||
BC103694.1 | - | 3, 4 | 655 | 40003751 |