Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR9A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001658.1 | NP_001001658.1 | 1 | 69 | ||
GenBank mRNA | BC137048.1 | - | 1 | 132 | 9021045 | |
BC137049.1 | - | 1 | 132 | 9021046 |
Set Membership: |
G-protein coupled receptor |