Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242773.2 | NP_001229702.1 | 3, 4 | 435 | ||
NM_001242774.2 | NP_001229703.1 | 4, 5 | 546 | |||
NM_001242775.2 | NP_001229704.1 | 3, 4 | 459 | |||
NM_001282876.1 | NP_001269805.1 | 2, 3 | 757 | |||
NM_001282877.1 | NP_001269806.1 | 3, 4 | 438 | |||
NM_153345.3 | NP_699176.1 | 2, 3 | 337 | |||
GenBank mRNA | AK075067.1 | - | 2, 3 | 282 | ||
AK172816.1 | - | 4, 5 | 488 | |||
AK315170.1 | - | 3, 4 | 383 | |||
AY358781.1 | - | 2, 3 | 352 | |||
BC035517.1 | - | 2, 3 | 737 | 5207904 | ||
DA626625.1 | - | 3, 4 | 398 | |||
HY180523.1 | - | 3, 4 | 439 |