Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242773.2 | NP_001229702.1 | 4 | 802 | ||
NM_001242774.2 | NP_001229703.1 | 5 | 913 | |||
NM_001242775.2 | NP_001229704.1 | 4 | 826 | |||
NM_001282876.1 | NP_001269805.1 | 3 | 1124 | |||
NM_001282877.1 | NP_001269806.1 | 4 | 805 | |||
NM_153345.3 | NP_699176.1 | 3 | 704 | |||
NR_040003.2 | 2 | 862 | ||||
NR_104250.1 | 3 | 543 | ||||
NR_104251.1 | 2 | 442 | ||||
NR_104252.1 | 3 | 540 | ||||
NR_104253.1 | 2 | 466 | ||||
NR_104254.1 | 3 | 564 | ||||
GenBank mRNA | AK075067.1 | - | 3 | 649 | ||
AK315170.1 | - | 4 | 750 | |||
AY358781.1 | - | 3 | 719 | |||
BC016719.1 | - | 1 | 74 | |||
BC035517.1 | - | 3 | 1104 | 5207904 | ||
BC035888.1 | - | 2 | 465 | |||
BE562464.1 | - | 2 | 449 | |||
BF036500.1 | - | 1 | 362 | |||
BI761527.1 | - | 2 | 392 | |||
CB156055.1 | - | 2 | 382 | |||
DB222115.1 | - | 3 | 483 | |||
DT220400.1 | - | 2 | 394 |