Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242773.2 | NP_001229702.1 | 3 | 278 | ||
NM_001242774.2 | NP_001229703.1 | 4 | 389 | |||
NM_001242775.2 | NP_001229704.1 | 3 | 302 | |||
NM_001282876.1 | NP_001269805.1 | 2 | 600 | |||
NM_001282877.1 | NP_001269806.1 | 3 | 281 | |||
NM_153345.3 | NP_699176.1 | 2 | 180 | |||
GenBank mRNA | AK027352.1 | - | 1 | 1090 | ||
AK075067.1 | - | 2 | 125 | |||
AK172816.1 | - | 4 | 331 | |||
AK315170.1 | - | 3 | 226 | |||
AY358781.1 | - | 2 | 195 | |||
BC035517.1 | - | 2 | 580 | 5207904 | ||
DA626625.1 | - | 3 | 241 | |||
HY180523.1 | - | 3 | 282 |