Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C7orf31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138811.3 | NP_620166.3 | 9 | 1479 | ||
XM_005249619.3 | XP_005249676.1 | 9 | 1328 | |||
XM_005249620.3 | XP_005249677.1 | 10 | 1859 | |||
XM_011515125.2 | XP_011513427.1 | 5 | 370 | |||
GenBank mRNA | AK097248.1 | - | 10 | 1200 | ||
AK098189.1 | - | 4 | 390 | |||
BC015397.1 | - | 2 | 209 | |||
BC043269.2 | - | 9 | 1582 | 5297014 | ||
BC060879.1 | - | 9 | 1392 | 5298874 | ||
BX640851.1 | - | 6 | 549 |