Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM92A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001283034.1 | NP_001269963.1 | 7, 8 | 812 | ||
NM_145269.4 | NP_660312.2 | 8, 9 | 926 | |||
NR_104268.1 | 9, 10 | 1012 | ||||
XM_005250787.2 | XP_005250844.1 | 8, 9 | 796 | |||
GenBank mRNA | AK096298.1 | - | 9, 10 | 906 | ||
BC108707.1 | - | 8, 9 | 888 | |||
CR627475.1 | - | 8, 9 | 1965 | |||
DQ327715.1 | - | 7, 8 | 726 | |||
DQ327716.1 | - | 8, 9 | 840 |