Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145153.1 | NP_001138625.1 | 7 | 839 | ||
NM_144649.2 | NP_653250.2 | 8 | 971 | |||
GenBank mRNA | AK057631.1 | - | 8 | 885 | ||
BC022053.2 | - | 4 | 487 | |||
BC062592.1 | - | 8 | 866 | 5224669 | ||
BC105783.1 | - | 7 | 757 | 4669222 |