Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145153.1 | NP_001138625.1 | 4 | 409 | ||
NM_144649.2 | NP_653250.2 | 4 | 409 | |||
GenBank mRNA | AK057631.1 | - | 4 | 323 | ||
BC062592.1 | - | 4 | 304 | 5224669 | ||
BC105783.1 | - | 4 | 327 | 4669222 | ||
BG505962.1 | - | 4 | 410 |