Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144649.2 | NP_653250.2 | 6 | 837 | ||
GenBank mRNA | AK057631.1 | - | 6 | 751 | ||
BC022053.2 | - | 2 | 353 | |||
BC062592.1 | - | 6 | 732 | 5224669 |