Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PXDNL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144651.4 | NP_653252.3 | 14, 15 | 1903 | ||
XM_006716420.3 | XP_006716483.1 | 12, 13 | 3201 | |||
XM_011517456.2 | XP_011515758.1 | 13, 14 | 3417 | |||
XM_011517457.2 | XP_011515759.1 | 13, 14 | 3417 | |||
XM_011517458.2 | XP_011515760.1 | 12, 13 | 1577 | |||
XM_011517459.2 | XP_011515761.1 | 14, 15 | 3489 | |||
XM_017013041.1 | XP_016868530.1 | 11, 12 | 1538 | |||
GenBank mRNA | AY877349.1 | - | 14, 15 | 1903 | ||
EU170240.1 | - | 14, 15 | 1802 | |||
KJ545488.1 | - | 12, 13 | 1614 |