Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RALYL SIRNA Assays ›
Gene Symbol:
RALYL
Gene Name:
RALY RNA binding protein-like
Gene Aliases:
HNRPCL3
Chromosome Location:
Chr.8: 84182629 - 84921844 on Build GRCh38
UniGene ID:
Hs.121663
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s44081
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RALYL
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001100391.1
NP_001093861.1
2
333
NM_001100392.1
NP_001093862.1
3
576
NM_001100393.1
NP_001093863.1
2
544
NM_001287243.1
NP_001274172.1
2
1257
NM_173848.5
NP_776247.3
2
1257
XM_011517462.2
XP_011515764.1
3
1361
XM_017013058.1
XP_016868547.1
2
532
XM_017013059.1
XP_016868548.1
4
627
XM_017013060.1
XP_016868549.1
2
2777
XM_017013061.1
XP_016868550.1
2
1929
XM_017013062.1
XP_016868551.1
4
736
XM_017013063.1
XP_016868552.1
2
499
XM_017013064.1
XP_016868553.1
3
850
XM_017013065.1
XP_016868554.1
2
398
XM_017013066.1
XP_016868555.1
2
544
XM_017013067.1
XP_016868556.1
3
1435
XM_017013068.1
XP_016868557.1
4
627
XM_017013069.1
XP_016868558.1
4
683
XM_017013070.1
XP_016868559.1
3
527
XM_017013071.1
XP_016868560.1
2
398
XM_017013072.1
XP_016868561.1
2
485
XM_017013073.1
XP_016868562.1
2
485
XM_017013074.1
XP_016868563.1
2
398
XM_017013075.1
XP_016868564.1
2
483
XM_017013076.1
XP_016868565.1
2
278
XM_017013077.1
XP_016868566.1
3
1528
XM_017013078.1
XP_016868567.1
2
544
XM_017013079.1
XP_016868568.1
3
527
XM_017013080.1
XP_016868569.1
2
278
XM_017013081.1
XP_016868570.1
2
278
XM_017013082.1
XP_016868571.1
2
544
GenBank mRNA
AF495529.1
-
2
1257
AF495530.1
-
2
474
AK094543.1
-
2
488
AK095026.1
-
2
1073
AK095560.1
-
3
527
BC031090.1
-
2
322
5311935
DA143105.1
-
2
544
DN989439.1
-
2
338
Back To Top
More Information
Gene Ontology Categories:
Process(es)
nucleotide binding
protein binding
identical protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-wjdsd:80/100.66.76.85:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline