Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCN8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178469.3 | NP_848564.2 | 2 | 479 | ||
XM_017014272.1 | XP_016869761.1 | 2 | 207 | |||
GenBank mRNA | AK090647.1 | - | 1 | 476 | ||
AK126902.1 | - | 1 | 1217 | |||
AY301268.1 | - | 2 | 199 | |||
BC130465.1 | - | 2 | 434 | 40146496 | ||
BC132714.1 | - | 2 | 434 | 40146736 |