Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM69B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152421.3 | NP_689634.2 | 5 | 1619 | ||
XM_006716954.3 | XP_006717017.1 | 5 | 1624 | |||
XM_011518238.2 | XP_011516540.1 | 6 | 1750 | |||
XM_017014273.1 | XP_016869762.1 | 4 | 1657 | |||
GenBank mRNA | AF318355.1 | - | 3 | 2362 | ||
AJ420457.1 | - | 4 | 1459 | |||
BC032097.2 | - | 5 | 1613 | 3161575 |