Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTRH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002913.1 | NP_001002913.1 | 4 | 453 | ||
XM_006716955.3 | XP_006717018.1 | 4 | 1510 | |||
XM_017014276.1 | XP_016869765.1 | 4 | 1510 | |||
XM_017014277.1 | XP_016869766.1 | 4 | 1510 | |||
XM_017014278.1 | XP_016869767.1 | 3 | 1410 | |||
GenBank mRNA | AK090922.1 | - | 5 | 1892 | ||
BC047012.1 | - | 4 | 453 | 5001221 |