Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253829.1 | NP_001240758.1 | 6 | 895 | ||
NM_001253830.1 | NP_001240759.1 | 7 | 956 | |||
NM_152422.4 | NP_689635.3 | 6 | 889 | |||
NM_177995.2 | NP_818931.1 | 7 | 971 | |||
XM_005251711.4 | XP_005251768.1 | 7 | 2913 | |||
XM_011518240.2 | XP_011516542.1 | 7 | 721 | |||
XM_011518241.2 | XP_011516543.1 | 7 | 675 | |||
XM_017014280.1 | XP_016869769.1 | 8 | 780 | |||
XM_017014281.1 | XP_016869770.1 | 7 | 699 | |||
XM_017014282.1 | XP_016869771.1 | 4 | 802 | |||
GenBank mRNA | AK124912.1 | - | 6 | 775 | ||
AK289692.1 | - | 6 | 846 | |||
AK310403.1 | - | 6 | 853 | |||
AY171233.1 | - | 7 | 976 | |||
BC067120.1 | - | 7 | 956 | 5539182 |