Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPP25L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148178.2 | NP_680544.1 | 2 | 342 | ||
NM_148179.2 | NP_680545.1 | 2 | 546 | |||
GenBank mRNA | AL528391.3 | - | 2 | 342 | ||
BC032136.2 | - | 2 | 308 | 4869926 | ||
BQ224958.1 | - | 2 | 510 | |||
DT218980.1 | - | 2 | 514 |