Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C9orf131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040410.2 | NP_001035500.1 | 2 | 284 | ||
NM_001040411.2 | NP_001035501.1 | 2 | 173 | |||
NM_001040412.2 | NP_001035502.1 | 3 | 248 | |||
NM_001287391.1 | NP_001274320.1 | 2 | 205 | |||
NM_203299.3 | NP_976044.2 | 2 | 389 | |||
GenBank mRNA | AK097595.1 | - | 2 | 355 | ||
BC045643.1 | - | 2 | 368 | 5298387 | ||
DB068600.1 | - | 2 | 389 | |||
DB072581.1 | - | 2 | 175 | |||
DB072610.1 | - | 2 | 175 | |||
DB077276.1 | - | 2 | 206 | |||
DB081756.1 | - | 2 | 167 | |||
DB091135.1 | - | 2 | 279 | |||
DB093341.1 | - | 3 | 213 | |||
DB096349.1 | - | 3 | 247 | |||
DB097641.1 | - | 1 | 124 |