Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLITRK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184749.2 | NP_001171678.1 | 2 | 981 | ||
NM_001184750.2 | NP_001171679.1 | 2 | 784 | |||
NM_173078.4 | NP_775101.1 | 2 | 934 | |||
XM_005262365.4 | XP_005262422.1 | 2 | 734 | |||
XM_011531265.2 | XP_011529567.1 | 2 | 779 | |||
GenBank mRNA | AK092016.1 | - | 2 | 651 | ||
AL713693.1 | - | 1 | 114 | |||
BC040986.1 | - | 2 | 943 | 4811695 |