Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PASD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173493.2 | NP_775764.2 | 14 | 2134 | ||
XM_011531102.1 | XP_011529404.1 | 14 | 2044 | |||
GenBank mRNA | AK097552.1 | - | 14 | 2047 | ||
AY270020.1 | - | 14 | 1940 | |||
AY623425.1 | - | 2 | 398 | |||
BC040301.1 | - | 14 | 1945 | 4826573 |