Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM199X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207318.3 | NP_997201.1 | 3, 4 | 886 | ||
XM_005262079.2 | XP_005262136.1 | 3, 4 | 883 | |||
XM_017029270.1 | XP_016884759.1 | 3, 4 | 455 | |||
GenBank mRNA | AK091221.1 | - | 4, 5 | 733 | ||
BC016683.1 | - | 1, 2 | 106 | |||
BC017665.1 | - | 3, 4 | 741 | 3903800 | ||
BC035886.1 | - | 2, 3 | 277 | |||
BC057847.1 | - | 3, 4 | 886 | 30344004 |