Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUNDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173794.3 | NP_776155.1 | 4 | 447 | ||
GenBank mRNA | BC035015.2 | - | 4 | 331 | 4812334 | |
BC042813.1 | - | 4 | 442 | 5274638 | ||
BM837991.1 | - | 4 | 473 |