Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081573.2 | NP_001075042.1 | 3 | 549 | ||
NM_001282283.1 | NP_001269212.1 | 3 | 549 | |||
NM_080612.3 | NP_542179.1 | 3 | 546 | |||
NR_104114.1 | 3 | 511 | ||||
XM_005274648.1 | XP_005274705.1 | 3 | 516 | |||
XM_006724804.1 | XP_006724867.1 | 3 | 516 | |||
XM_011531103.1 | XP_011529405.1 | 3 | 516 | |||
XM_011531105.1 | XP_011529407.1 | 3 | 516 | |||
XM_011531106.1 | XP_011529408.1 | 3 | 516 | |||
XM_017029276.1 | XP_016884765.1 | 3 | 516 | |||
GenBank mRNA | AK126283.1 | - | 3 | 514 | ||
AK307927.1 | - | 3 | 543 | |||
AL832051.1 | - | 3 | 470 | |||
AY057989.1 | - | 3 | 520 | |||
CN481476.1 | - | 3 | 509 | |||
DB065120.1 | - | 3 | 508 |