Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAGE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017931.2 | NP_001017931.2 | 5 | 648 | ||
NM_001171252.1 | NP_001164723.1 | 5 | 662 | |||
NM_001303613.1 | NP_001290542.1 | 5 | 450 | |||
XM_017029282.1 | XP_016884771.1 | 5 | 451 | |||
GenBank mRNA | BC140001.1 | - | 5 | 521 | 8860308 | |
BC140741.1 | - | 5 | 534 | 9021609 | ||
BC144449.1 | - | 5 | 547 | 9052975 |