Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083615.3 | NP_001077084.2 | 18 | 2229 | ||
NM_138995.4 | NP_620482.3 | 18 | 2229 | |||
NR_045682.1 | 18 | 2229 | ||||
NR_045683.1 | 18 | 2229 | ||||
NR_045684.1 | 18 | 2229 | ||||
XM_006712299.3 | XP_006712362.1 | 18 | 2117 | |||
XM_011510654.2 | XP_011508956.1 | 18 | 2117 | |||
XM_011510655.2 | XP_011508957.1 | 18 | 2117 | |||
XM_011510656.2 | XP_011508958.1 | 12 | 1492 | |||
XM_011510657.2 | XP_011508959.1 | 11 | 1364 | |||
XM_011510658.2 | XP_011508960.1 | 12 | 1673 | |||
GenBank mRNA | AF369908.1 | - | 18 | 2229 | ||
AF391554.1 | - | 18 | 2229 | |||
AF391555.1 | - | 18 | 2229 | |||
AF391556.1 | - | 18 | 2229 | |||
AF391557.1 | - | 18 | 2229 | |||
AF391558.1 | - | 18 | 2229 | |||
AK055778.1 | - | 18 | 2117 | |||
BC136620.1 | - | 18 | 2166 | 9020610 | ||
BC144307.1 | - | 18 | 2166 | 9052827 | ||
BX647895.1 | - | 16 | 1954 |