Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHODL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204174.1 | NP_001191103.1 | 4, 5 | 658 | ||
NM_001204175.1 | NP_001191104.1 | 4, 5 | 624 | |||
NM_001204176.1 | NP_001191105.1 | 5, 6 | 724 | |||
NM_024944.2 | NP_079220.2 | 4, 5 | 1033 | |||
XM_011529457.2 | XP_011527759.1 | 5, 6 | 1030 | |||
GenBank mRNA | AF257472.1 | - | 4, 5 | 1033 | ||
AF523314.1 | - | 5, 6 | 724 | |||
AK022689.1 | - | 5, 6 | 906 | |||
AK296009.1 | - | 4, 5 | 658 | |||
AK313725.1 | - | 4, 5 | 681 | |||
AY358608.1 | - | 4, 5 | 1019 | |||
BC009418.1 | - | 4, 5 | 1003 | 3509743 |