Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080430.2 | NP_536355.1 | 5 | 358 | ||
GenBank mRNA | AY043487.1 | - | 5 | 387 | ||
BC013421.1 | - | 5 | 304 | 3890282 | ||
BC030236.1 | - | 5 | 315 | 4184342 | ||
BC068004.1 | - | 5 | 348 | 6058758 |