Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133494.2 | NP_598001.1 | 7 | 883 | ||
XM_017000344.1 | XP_016855833.1 | 7 | 885 | |||
XM_017000345.1 | XP_016855834.1 | 8 | 992 | |||
XM_017000346.1 | XP_016855835.1 | 7 | 885 | |||
XM_017000347.1 | XP_016855836.1 | 7 | 885 | |||
XM_017000348.1 | XP_016855837.1 | 7 | 885 | |||
GenBank mRNA | AB062450.1 | - | 6 | 602 | ||
AK313411.1 | - | 7 | 683 |