Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C20orf96 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK091924.1 | - | 6 | 749 | ||
AK097394.1 | - | 8 | 910 | |||
AK302223.1 | - | 6 | 657 | |||
AK304526.1 | - | 6 | 764 | |||
BC137450.1 | - | 8 | 906 | 9021450 | ||
BC144648.1 | - | 7 | 857 | |||
BC144649.1 | - | 8 | 910 | 9053180 | ||
BX640715.1 | - | 7 | 883 |