Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPIFA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319164.1 | NP_001306093.1 | 2 | 163 | ||
NM_080574.3 | NP_542141.1 | 2 | 245 | |||
GenBank mRNA | AF432917.1 | - | 1 | 97 | ||
AK291854.1 | - | 2 | 245 | |||
AY359055.1 | - | 2 | 163 | |||
BC065726.1 | - | 2 | 172 | 30326122 |