Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025224.3 | NP_079500.2 | 4 | 1438 | ||
XM_005260195.4 | XP_005260252.1 | 4 | 1518 | |||
XM_005260196.3 | XP_005260253.1 | 4 | 1334 | |||
XM_005260197.4 | XP_005260254.1 | 4 | 1654 | |||
XM_005260198.4 | XP_005260255.1 | 4 | 1317 | |||
XM_006723700.3 | XP_006723763.1 | 4 | 1350 | |||
XM_011528548.2 | XP_011526850.1 | 4 | 1618 | |||
GenBank mRNA | AK126720.1 | - | 3 | 455 | ||
AK131482.1 | - | 4 | 1438 | |||
BC052269.1 | - | 4 | 1418 | 6574149 | ||
BC073800.1 | - | 4 | 1418 | 6576441 |