Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WFDC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080614.1 | NP_542181.1 | 4 | 421 | ||
XM_011528553.2 | XP_011526855.1 | 4 | 431 | |||
GenBank mRNA | AF488306.1 | - | 4 | 421 | ||
AL591713.1 | - | 3 | 318 | |||
BC026014.1 | - | 3 | 325 | |||
BC102013.1 | - | 3 | 327 | |||
BC102014.1 | - | 2 | 198 |