Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOL4L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256798.1 | NP_001243727.1 | 7, 8 | 1496 | ||
NM_080616.4 | NP_542183.2 | 4, 5 | 835 | |||
XM_005260282.4 | XP_005260339.1 | 7, 8 | 1782 | |||
XM_005260283.4 | XP_005260340.1 | 8, 9 | 1806 | |||
XM_005260284.4 | XP_005260341.1 | 7, 8 | 1081 | |||
XM_005260285.4 | XP_005260342.2 | 7, 8 | 1786 | |||
XM_005260286.4 | XP_005260343.1 | 7, 8 | 1706 | |||
XM_005260288.2 | XP_005260345.1 | 4, 5 | 883 | |||
XM_005260289.4 | XP_005260346.1 | 4, 5 | 834 | |||
XM_006723702.3 | XP_006723765.1 | 8, 9 | 1173 | |||
XM_006723703.3 | XP_006723766.1 | 7, 8 | 1085 | |||
XM_011528562.2 | XP_011526864.1 | 7, 8 | 1490 | |||
XM_011528563.1 | XP_011526865.1 | 7, 8 | 1490 | |||
XM_017027669.1 | XP_016883158.1 | 9, 10 | 1827 | |||
XM_017027670.1 | XP_016883159.1 | 4, 5 | 738 | |||
GenBank mRNA | AK056286.1 | - | 4, 5 | 638 | ||
BC065370.1 | - | 3, 4 | 607 | 6261963 |