Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MACROD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033087.1 | NP_001028259.1 | 10 | 756 | ||
NM_080676.5 | NP_542407.2 | 14 | 1263 | |||
XM_017027675.1 | XP_016883164.1 | 7 | 400 | |||
XM_017027676.1 | XP_016883165.1 | 7 | 400 | |||
XM_017027677.1 | XP_016883166.1 | 14 | 1287 | |||
GenBank mRNA | AK125899.1 | - | 7 | 490 | ||
AK131348.1 | - | 14 | 1393 | |||
BC018687.1 | - | 10 | 759 | 4717477 | ||
BC035876.1 | - | 2 | 200 | |||
EF159161.1 | - | 999 |