Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYNLL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080677.2 | NP_542408.1 | 3 | 695 | ||
GenBank mRNA | AF086304.1 | - | 2 | 403 | ||
AF112997.1 | - | 2 | 675 | |||
BC010744.1 | - | 3 | 684 | 3891655 | ||
BF528423.1 | - | 3 | 687 |