Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080722.4 | NP_542453.2 | 7 | 1480 | ||
NM_139155.3 | NP_631894.2 | 7 | 1489 | |||
XM_011539300.2 | XP_011537602.1 | 7 | 1473 | |||
XM_011539301.2 | XP_011537603.1 | 4 | 486 | |||
XM_011539302.2 | XP_011537604.1 | 4 | 363 | |||
XM_011539303.2 | XP_011537605.1 | 7 | 1477 | |||
XM_011539305.2 | XP_011537607.1 | 7 | 1477 | |||
XM_011539306.2 | XP_011537608.1 | 7 | 1477 | |||
XM_011539307.2 | XP_011537609.1 | 7 | 1477 | |||
XM_011539308.2 | XP_011537610.1 | 7 | 1477 | |||
GenBank mRNA | AF358666.1 | - | 7 | 1183 | ||
AF366351.1 | - | 6 | 1110 | |||
AJ345098.1 | - | 7 | 1183 |