Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCP2D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178483.2 | NP_848578.1 | 1 | 274 | ||
GenBank mRNA | AF418568.1 | - | 1 | 274 | ||
AF418570.1 | - | 1 | 274 | |||
AK130970.1 | - | 1 | 270 | |||
AY248902.1 | - | 1 | 231 | |||
BC105121.1 | - | 1 | 216 | 8144124 | ||
BC112024.1 | - | 1 | 216 | 8327492 |