Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WFDC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080827.1 | NP_543017.1 | 2 | 211 | ||
GenBank mRNA | AF411861.1 | - | 2 | 211 | ||
BC101600.1 | - | 2 | 206 | 8069106 | ||
BC101604.1 | - | 2 | 206 | 8069110 | ||
GQ472217.1 | - | 2 | 206 |
Gene Symbol: EPPIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DC395362.1 | - | 4 | 493 |
Gene Symbol: EPPIN-WFDC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198986.1 | NP_001185915.1 | 4 | 492 | ||
GenBank mRNA | AK301937.1 | - | 4 | 467 |