Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEFB129 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080831.3 | NP_543021.1 | 2 | 308 | ||
GenBank mRNA | AF525931.1 | - | 2 | 277 | ||
AF529413.1 | - | 2 | 277 | |||
AY122480.1 | - | 2 | 282 | |||
AY358186.1 | - | 2 | 282 | |||
BC070359.1 | - | 2 | 308 | 6592149 |