Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258315.1 | NP_001245244.1 | 14 | 1672 | ||
NM_001258316.1 | NP_001245245.1 | 14 | 1902 | |||
NM_018098.5 | NP_060568.3 | 13 | 1579 | |||
XM_005247176.1 | XP_005247233.1 | 13 | 1582 | |||
XM_006713523.3 | XP_006713586.1 | 14 | 1661 | |||
XM_006713524.3 | XP_006713587.1 | 14 | 1659 | |||
XM_011512514.2 | XP_011510816.1 | 15 | 1985 | |||
XM_011512515.2 | XP_011510817.1 | 14 | 1641 | |||
XM_011512516.1 | XP_011510818.1 | 13 | 1543 | |||
XM_017005830.1 | XP_016861319.1 | 15 | 1949 | |||
XM_017005831.1 | XP_016861320.1 | 14 | 1576 | |||
XM_017005832.1 | XP_016861321.1 | 14 | 1890 | |||
XM_017005833.1 | XP_016861322.1 | 14 | 1876 | |||
XM_017005834.1 | XP_016861323.1 | 15 | 1982 | |||
XM_017005835.1 | XP_016861324.1 | 15 | 1923 | |||
XM_017005836.1 | XP_016861325.1 | 14 | 1610 | |||
XM_017005837.1 | XP_016861326.1 | 13 | 1495 | |||
XM_017005838.1 | XP_016861327.1 | 13 | 1456 | |||
GenBank mRNA | AK023267.1 | - | 12 | 1412 | ||
AK027713.1 | - | 3 | 336 | |||
AK308391.1 | - | 14 | 1594 | |||
AK314581.1 | - | 13 | 1523 | |||
AY376439.1 | - | 14 | 1825 | |||
BC112086.1 | - | 12 | 1458 | 8327554 | ||
DQ847274.1 | - | 13 | 1477 |