Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBBP8NL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080833.2 | NP_543023.2 | 5, 6 | 519 | ||
GenBank mRNA | AK074919.1 | - | 5, 6 | 479 | ||
AK290299.1 | - | 5, 6 | 492 | |||
BC137326.1 | - | 4, 5 | 407 | 9021324 | ||
BC137327.1 | - | 4, 5 | 407 | 9021325 | ||
DA871775.1 | - | 5, 6 | 521 |