Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080836.3 | NP_543026.2 | 3 | 1016 | ||
GenBank mRNA | AF195026.1 | - | 2 | 617 | ||
AK314728.1 | - | 3 | 868 | |||
BC017340.2 | - | 2 | 599 | |||
BC111573.1 | - | 2 | 156 | |||
BC140883.1 | - | 2 | 717 | 9021751 | ||
BC140884.1 | - | 2 | 717 | 9021752 | ||
GQ281297.1 | - | 3 | 1016 |