Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC34A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177316.1 | NP_001170787.1 | 6, 7 | 775 | ||
NM_001177317.1 | NP_001170788.1 | 6, 7 | 778 | |||
NM_080877.2 | NP_543153.1 | 6, 7 | 740 | |||
XM_011518257.2 | XP_011516559.1 | 6, 7 | 716 | |||
XM_011518258.1 | XP_011516560.1 | 5, 6 | 606 | |||
XM_011518259.1 | XP_011516561.1 | 6, 7 | 740 | |||
XM_011518260.1 | XP_011516562.1 | 6, 7 | 658 | |||
XM_011518261.2 | XP_011516563.1 | 6, 7 | 731 | |||
XM_017014290.1 | XP_016869779.1 | 7, 8 | 826 | |||
XM_017014291.1 | XP_016869780.1 | 6, 7 | 716 | |||
XM_017014292.1 | XP_016869781.1 | 6, 7 | 716 | |||
GenBank mRNA | AB055000.1 | - | 6, 7 | 624 | ||
AK095999.1 | - | 6, 7 | 740 | |||
AL832449.1 | - | 6, 7 | 1786 | |||
BC029942.1 | - | 6, 7 | 778 |