Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130388.3 | NP_569059.3 | 2 | 890 | ||
GenBank mRNA | AF403030.1 | - | 1 | 670 | ||
AK096896.1 | - | 2 | 870 | |||
BC069436.1 | - | 1 | 670 | 7262244 | ||
BC069555.1 | - | 1 | 670 | 7262256 | ||
BC093883.1 | - | 1 | 731 | 7939728 | ||
BC104946.1 | - | 1 | 731 | 8143949 |