Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101929926 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322244.1 | NP_001309173.1 | 2, 3 | 251 | ||
NR_136239.1 | 2, 3 | 251 | ||||
XM_011512269.1 | XP_011510571.1 | 2, 3 | 313 | |||
XM_011512270.1 | XP_011510572.1 | 2, 3 | 313 | |||
XM_011512271.1 | XP_011510573.1 | 2, 3 | 259 | |||
XM_011512272.1 | XP_011510574.1 | 2, 3 | 313 | |||
XM_011512273.1 | XP_011510575.1 | 2, 3 | 313 | |||
XM_011512274.1 | XP_011510576.1 | 2, 3 | 313 | |||
XM_011512275.1 | XP_011510577.1 | 2, 3 | 313 | |||
XM_011512276.1 | XP_011510578.1 | 2, 3 | 313 | |||
XM_011512277.2 | XP_011510579.1 | 2, 3 | 313 | |||
XM_011512278.1 | XP_011510580.1 | 2, 3 | 313 | |||
XM_011512279.1 | XP_011510581.1 | 2, 3 | 313 | |||
XM_017003148.1 | XP_016858637.1 | 2, 3 | 313 | |||
XM_017003149.1 | XP_016858638.1 | 2, 3 | 313 | |||
XM_017003150.1 | XP_016858639.1 | 2, 3 | 259 | |||
GenBank mRNA | BI828150.1 | - | 2, 3 | 220 | ||
BX113116.1 | - | 2, 3 | 229 |