Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C10orf82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144661.2 | NP_653262.1 | 5 | 781 | ||
XM_011539344.2 | XP_011537646.1 | 6 | 1175 | |||
XM_011539345.2 | XP_011537647.1 | 6 | 1179 | |||
XM_011539346.2 | XP_011537648.1 | 6 | 1179 | |||
XM_011539347.2 | XP_011537649.1 | 5 | 1218 | |||
XM_017015753.1 | XP_016871242.1 | 6 | 796 | |||
XM_017015754.1 | XP_016871243.1 | 6 | 922 | |||
XM_017015755.1 | XP_016871244.1 | 6 | 938 | |||
GenBank mRNA | AK097587.1 | - | 5 | 1408 | ||
BC021737.2 | - | 5 | 781 | 4821913 |