Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135091.1 | NP_001128563.1 | 3 | 931 | ||
NM_001135092.1 | NP_001128564.1 | 3 | 931 | |||
NM_145650.3 | NP_663625.2 | 2 | 843 | |||
XM_017017235.1 | XP_016872724.1 | 3 | 931 | |||
XM_017017236.1 | XP_016872725.1 | 3 | 931 | |||
GenBank mRNA | AJ417818.1 | - | 3 | 799 | ||
AJ507429.1 | - | 3 | 805 | |||
AK128337.1 | - | 2 | 696 | |||
AY358668.1 | - | 3 | 896 | |||
BC020912.1 | - | 3 | 923 | 4733074 | ||
BC058007.1 | - | 3 | 809 | 4719841 |