Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIWIL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152431.2 | NP_689644.2 | 2 | 314 | ||
GenBank mRNA | AB079366.1 | - | 2 | 274 | ||
AK093475.1 | - | 2 | 254 | |||
AK293616.1 | - | 2 | 166 | |||
BC031060.1 | - | 2 | 106 | 5271278 | ||
CR749642.1 | - | 2 | 242 | |||
DB091271.1 | - | 2 | 314 |