Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CWF19L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152434.2 | NP_689647.2 | 12, 13 | 1902 | ||
XM_017017239.1 | XP_016872728.1 | 11, 12 | 1220 | |||
GenBank mRNA | AK056905.1 | - | 10, 11 | 1554 | ||
AK126881.1 | - | 12, 13 | 1893 | |||
AK293001.1 | - | 12, 13 | 1837 | |||
BC110439.1 | - | 9, 10 | 1515 | 40001346 | ||
BC110440.1 | - | 9, 10 | 1515 | 40001348 | ||
BC110442.1 | - | 9, 10 | 1515 | 40001350 | ||
BC118669.1 | - | 9, 10 | 1518 | 40012545 | ||
BX647832.1 | - | 12, 13 | 1876 | |||
BX648704.1 | - | 12, 13 | 1881 | |||
BX649022.1 | - | 12, 13 | 1872 |